Assay Details
Gene Symbol: Irf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106108.1 | NP_001099578.1 | 6 - 7 | 810 | 78 | |
XM_006253899.2 | XP_006253961.1 | 6 - 7 | 882 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
