Assay Details
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031029.1 | NP_112291.1 | 6 - 7 | 724 | 70 | |
XM_008767594.1 | XP_008765816.1 | 7 - 8 | 1310 | 70 | ||
XM_008767595.1 | XP_008765817.1 | 7 - 8 | 1316 | 70 | ||
GenBank mRNA | FQ222085.1 | - | 7 - 8 | 891 | 70 | |
U95368.1 | - | 6 - 7 | 724 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
