Assay Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109190.2 | NP_001102660.1 | 1 - 2 | 919 | 77 | |
XM_006232897.2 | XP_006232959.1 | 1 - 2 | 845 | 77 | ||
XM_006232898.2 | XP_006232960.1 | 1 - 2 | 846 | 77 | ||
XM_006232900.2 | XP_006232962.1 | 1 - 2 | 845 | 77 | ||
XM_006232902.2 | XP_006232964.1 | 1 - 2 | 845 | 77 | ||
XM_006232904.2 | XP_006232966.1 | 2 - 3 | 2573 | 77 | ||
XM_006232905.2 | XP_006232967.1 | 2 - 3 | 2606 | 77 | ||
XM_006232908.2 | XP_006232970.1 | 1 - 2 | 843 | 77 | ||
XM_008761322.1 | XP_008759544.1 | 2 - 3 | 3160 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232904.2, XM_006232905.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
