Assay Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109190.2 | NP_001102660.1 | 6 - 7 | 1398 | 67 | |
XM_006232897.2 | XP_006232959.1 | 6 - 7 | 1324 | 67 | ||
XM_006232898.2 | XP_006232960.1 | 6 - 7 | 1322 | 67 | ||
XM_006232900.2 | XP_006232962.1 | 6 - 7 | 1321 | 67 | ||
XM_006232902.2 | XP_006232964.1 | 6 - 7 | 1324 | 67 | ||
XM_006232903.2 | XP_006232965.1 | 5 - 6 | 1695 | 67 | ||
XM_006232904.2 | XP_006232966.1 | 7 - 8 | 3052 | 67 | ||
XM_006232905.2 | XP_006232967.1 | 7 - 8 | 3085 | 67 | ||
XM_006232906.2 | XP_006232968.1 | 5 - 6 | 4157 | 67 | ||
XM_006232907.2 | XP_006232969.1 | 5 - 6 | 642 | 67 | ||
XM_006232908.2 | XP_006232970.1 | 6 - 7 | 1322 | 67 | ||
XM_006232911.2 | XP_006232973.1 | 5 - 6 | 519 | 67 | ||
XM_006232912.2 | XP_006232974.1 | 4 - 5 | 441 | 67 | ||
XM_008761322.1 | XP_008759544.1 | 7 - 8 | 3639 | 67 | ||
XM_008761323.1 | XP_008759545.1 | 6 - 7 | 608 | 67 | ||
XM_008761324.1 | XP_008759546.1 | 5 - 6 | 2762 | 67 | ||
XM_008761325.1 | XP_008759547.1 | 5 - 6 | 2716 | 67 | ||
XM_008761326.1 | XP_008759548.1 | 5 - 6 | 2713 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
