Assay Details
Gene Symbol: Dmrtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109140.1 | NP_001102610.1 | 2 - 3 | 334 | 62 |
Gene Symbol: LOC100910371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244579.2 | XP_006244641.1 | 2 - 3 | 388 | 62 | |
XM_008766899.1 | XP_008765121.1 | 1 - 2 | 987 | 62 | ||
XM_008766900.1 | XP_008765122.1 | 1 - 2 | 987 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
