Assay Details
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012497.1 | NP_036629.1 | 1 - 2 | 98 | 63 | |
GenBank mRNA | BC099749.1 | - | 1 - 2 | 84 | 5599032 | 63 |
FQ213506.1 | - | 1 - 2 | 101 | 63 | ||
FQ214420.1 | - | 1 - 2 | 124 | 63 | ||
FQ228893.1 | - | 1 - 2 | 98 | 63 | ||
X06984.1 | - | 1 - 2 | 98 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC099749.1, NM_012497.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
