Assay Details
Gene Symbol: Cyp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012541.3 | NP_036673.3 | 6 - 7 | 1296 | 68 | |
GenBank mRNA | BC127476.1 | - | 6 - 7 | 1296 | 7373253 | 68 |
FQ209607.1 | - | 5 - 6 | 724 | 68 | ||
FQ210213.1 | - | 6 - 7 | 1302 | 68 | ||
FQ210219.1 | - | 5 - 6 | 444 | 68 | ||
K02422.1 | - | 5 - 6 | 1264 | 68 | ||
M26127.1 | - | 5 - 6 | 453 | 68 | ||
X01031.1 | - | 5 - 6 | 811 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
