Assay Details
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012573.3 | NP_036705.3 | 5 - 6 | 1548 | 59 | |
GenBank mRNA | D13211.1 | - | 4 - 5 | 1341 | 59 | |
M91561.1 | - | 5 - 6 | 1548 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
