Assay Details
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012573.3 | NP_036705.3 | 7 - 8 | 1872 | 63 | |
GenBank mRNA | D13211.1 | - | 6 - 7 | 1665 | 63 | |
M91561.1 | - | 7 - 8 | 1872 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
