Assay Details
Gene Symbol: Myc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012603.2 | NP_036735.2 | 1 - 2 | 562 | 84 | |
GenBank mRNA | AY679729.1 | - | 1 - 2 | 399 | 84 | |
AY679730.1 | - | 1 - 2 | 562 | 84 | ||
BC091699.1 | - | 1 - 2 | 368 | 7303974 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY679729.1, AY679730.1, BC091699.1, NM_012603.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
