Assay Details
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012624.3 | NP_036756.3 | 9 - 10 | 1358 | 69 | |
XM_006232593.2 | XP_006232655.1 | 9 - 10 | 1489 | 69 | ||
GenBank mRNA | BC161827.1 | - | 8 - 9 | 1348 | 69 | |
FQ211172.1 | - | 3 - 4 | 786 | 69 | ||
M11709.1 | - | 9 - 10 | 1359 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
