Assay Details
Gene Symbol: Prlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034111.1 | NP_001029283.1 | 5 - 6 | 553 | 124 | |
NM_012630.2 | NP_036762.1 | 5 - 6 | 563 | 124 | ||
GenBank mRNA | BC092133.1 | - | 5 - 6 | 557 | 124 | |
M19304.1 | - | 5 - 6 | 564 | 124 | ||
M34083.1 | - | 4 - 5 | 449 | 124 | ||
M57668.1 | - | 5 - 6 | 553 | 124 | ||
M74152.1 | - | 5 - 6 | 523 | 124 | ||
U07567.1 | - | 5 - 6 | 506 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
