Assay Details
Gene Symbol: Tac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124768.1 | NP_001118240.1 | 5 - 6 | 395 | 64 | |
NM_012666.2 | NP_036798.1 | 6 - 7 | 440 | 64 | ||
XM_008762692.1 | XP_008760914.1 | 6 - 7 | 581 | 64 | ||
XM_008762693.1 | XP_008760915.1 | 5 - 6 | 533 | 64 | ||
GenBank mRNA | M14312.1 | - | 4 - 5 | 307 | 64 | |
M15191.1 | - | 6 - 7 | 442 | 64 | ||
M34183.1 | - | 5 - 6 | 397 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
