Assay Details
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012762.2 | NP_036894.2 | 7 - 8 | 1108 | 74 | |
GenBank mRNA | D85899.1 | - | 7 - 8 | 1108 | 74 | |
FQ228818.1 | - | 5 - 6 | 741 | 74 | ||
U14647.1 | - | 7 - 8 | 1108 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
