Assay Details
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012881.2 | NP_037013.2 | 1 - 2 | 94 | 59 | |
GenBank mRNA | BC078874.1 | - | 1 - 2 | 74 | 7110677 | 59 |
CV102664.1 | - | 1 - 2 | 94 | 59 | ||
FQ213473.1 | - | 1 - 2 | 83 | 59 | ||
FQ222192.1 | - | 1 - 2 | 110 | 59 | ||
M14656.1 | - | 1 - 2 | 67 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC078874.1, M14656.1, NM_012881.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
