Assay Details
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012949.2 | NP_037081.2 | 7 - 8 | 717 | 67 | |
XM_006246599.2 | XP_006246661.1 | 7 - 8 | 788 | 67 | ||
XM_006246600.2 | XP_006246662.1 | 7 - 8 | 768 | 67 | ||
GenBank mRNA | BC083566.1 | - | 7 - 8 | 704 | 67 | |
FQ214810.1 | - | 6 - 7 | 695 | 67 | ||
FQ214945.1 | - | 7 - 8 | 735 | 67 | ||
FQ215034.1 | - | 3 - 4 | 394 | 67 | ||
FQ215526.1 | - | 7 - 8 | 727 | 67 | ||
FQ215648.1 | - | 7 - 8 | 727 | 67 | ||
FQ215759.1 | - | 7 - 8 | 739 | 67 | ||
FQ215865.1 | - | 7 - 8 | 727 | 67 | ||
FQ216150.1 | - | 6 - 7 | 695 | 67 | ||
FQ217095.1 | - | 7 - 8 | 727 | 67 | ||
FQ223831.1 | - | 3 - 4 | 394 | 67 | ||
FQ224938.1 | - | 7 - 8 | 727 | 67 | ||
Y00979.1 | - | 6 - 7 | 679 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
