Assay Details
Gene Symbol: Sirpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013016.2 | NP_037148.2 | 7 - 8 | 1465 | 86 | |
XM_006234951.2 | XP_006235013.1 | 6 - 7 | 1763 | 86 | ||
XM_006234952.2 | XP_006235014.1 | 7 - 8 | 2125 | 86 | ||
XM_006234953.2 | XP_006235015.1 | 7 - 8 | 2307 | 86 | ||
GenBank mRNA | BC099773.1 | - | 7 - 8 | 1401 | 86 | |
D38468.1 | - | 6 - 7 | 1520 | 86 | ||
D85183.1 | - | 6 - 7 | 1277 | 86 | ||
U62328.1 | - | 7 - 8 | 1465 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
