Assay Details
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013032.3 | NP_037164.3 | 8 - 9 | 954 | 100 | |
GenBank mRNA | BC061743.1 | - | 8 - 9 | 954 | 100 | |
D63772.1 | - | 985 | 100 | |||
U21104.1 | - | 8 - 9 | 966 | 100 | ||
U39555.1 | - | 8 - 9 | 1044 | 100 | ||
X94255.1 | - | 8 - 9 | 1011 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
