Assay Details
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013033.2 | NP_037165.1 | 1 - 2 | 307 | 73 | |
GenBank mRNA | AB000729.1 | - | 1 - 2 | 333 | 73 | |
BC081827.1 | - | 1 - 2 | 339 | 73 | ||
D16101.1 | - | 1 - 2 | 307 | 73 | ||
U03120.1 | - | 1 - 2 | 265 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
