Assay Details
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013059.1 | NP_037191.1 | 2 - 3 | 207 | 105 | |
XM_006239136.2 | XP_006239198.1 | 2 - 3 | 581 | 105 | ||
GenBank mRNA | BC088399.1 | - | 2 - 3 | 230 | 7128677 | 105 |
J03572.1 | - | 2 - 3 | 207 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC088399.1, J03572.1, NM_013059.1, XM_006239136.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
