Assay Details
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013091.1 | NP_037223.1 | 1 - 2 | 275 | 121 | |
XM_006237334.2 | XP_006237396.1 | 1 - 2 | 348 | 121 | ||
GenBank mRNA | AF329976.1 | - | 1 - 2 | 274 | 121 | |
AF329977.1 | - | 1 - 2 | 274 | 121 | ||
AF329978.1 | - | 1 - 2 | 274 | 121 | ||
AF329979.1 | - | 1 - 2 | 274 | 121 | ||
AF329980.1 | - | 1 - 2 | 274 | 121 | ||
AF329981.1 | - | 1 - 2 | 274 | 121 | ||
BC086413.1 | - | 1 - 2 | 295 | 121 | ||
M63122.1 | - | 1 - 2 | 275 | 121 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF329976.1, AF329977.1, AF329978.1, AF329979.1, AF329980.1, AF329981.1, BC086413.1, M63122.1, NM_013091.1, XM_006237334.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
