Assay Details
Gene Symbol: Dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013097.2 | NP_037229.1 | 2 - 3 | 423 | 70 | |
XM_006245829.2 | XP_006245891.1 | 3 - 4 | 1103 | 70 | ||
GenBank mRNA | AF397150.1 | - | 2 - 3 | 423 | 70 | |
AF397151.1 | - | 3 - 4 | 294 | 70 | ||
BC089764.1 | - | 2 - 3 | 423 | 70 | ||
X56060.1 | - | 2 - 3 | 276 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
