Assay Details
Gene Symbol: Adrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013108.2 | NP_037240.2 | 1 - 2 | 1383 | 58 | |
XM_008771315.1 | XP_008769537.1 | 1 - 2 | 1708 | 58 | ||
GenBank mRNA | M74716.1 | - | 1 - 2 | 1382 | 58 | |
S56481.1 | - | 1 - 2 | 1174 | 58 | ||
S73473.1 | - | 1 - 2 | 1213 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
