Assay Details
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013151.2 | NP_037283.2 | 9 - 10 | 993 | 109 | |
XM_006253347.2 | XP_006253409.1 | 9 - 10 | 1065 | 109 | ||
XM_008771348.1 | XP_008769570.1 | 10 - 11 | 1123 | 109 | ||
GenBank mRNA | BC061565.1 | - | 9 - 10 | 993 | 6920438 | 109 |
M23697.1 | - | 8 - 9 | 917 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
