Assay Details
Gene Symbol: Il2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013195.1 | NP_037327.1 | 1 - 2 | 83 | 82 | |
GenBank mRNA | M55050.1 | - | 1 - 2 | 83 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M55050.1, NM_013195.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
