Assay Details
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134350.2 | NP_599177.2 | 1 - 2 | 91 | 88 | |
XM_006248150.2 | XP_006248212.1 | 3 - 4 | 235 | 88 | ||
XM_008768568.1 | XP_008766790.1 | 3 - 4 | 169 | 88 | ||
GenBank mRNA | BC099808.1 | - | 1 - 2 | 91 | 7938638 | 88 |
X52712.1 | - | 1 - 2 | 166 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC099808.1, NM_134350.2, X52712.1, XM_006248150.2, XM_008768568.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
