Assay Details
Gene Symbol: Sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017051.2 | NP_058747.1 | 1 - 2 | 105 | 76 | |
GenBank mRNA | BC070913.1 | - | 1 - 2 | 105 | 7104698 | 76 |
Y00497.1 | - | 1 - 2 | 33 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC070913.1, NM_017051.2, Y00497.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
