Assay Details
Gene Symbol: Tf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013110.1 | NP_001013128.1 | 1 - 2 | 73 | 67 | |
GenBank mRNA | AF476964.1 | - | 1 - 2 | 104 | 67 | |
AY327504.1 | - | 1 - 2 | 42 | 67 | ||
BC087021.1 | - | 1 - 2 | 73 | 67 | ||
D38380.1 | - | 1 - 2 | 72 | 67 | ||
X77158.1 | - | 1 - 2 | 59 | 67 |
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY325214.1 | - | 82 | 67 | ||
AY325230.1 | - | 82 | 67 | |||
AY325241.1 | - | 82 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
