Assay Details
Gene Symbol: Cth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017074.1 | NP_058770.1 | 1 - 2 | 176 | 66 | |
GenBank mRNA | AB052882.2 | - | 1 - 2 | 175 | 66 | |
AY032875.1 | - | 1 - 2 | 175 | 66 | ||
AY641456.1 | - | 1 - 2 | 175 | 66 | ||
BC078869.1 | - | 1 - 2 | 189 | 7109746 | 66 | |
D17370.1 | - | 1 - 2 | 176 | 66 | ||
X53460.1 | - | 64 | 66 |
Gene Symbol: LOC103691744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761574.1 | XP_008759796.1 | 223 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
