Assay Details
Gene Symbol: Gnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017084.1 | NP_058780.1 | 1 - 2 | 213 | 107 | |
GenBank mRNA | X06150.1 | - | 1 - 2 | 213 | 107 |
Gene Symbol: LOC100911564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766920.1 | XP_008765142.1 | 1 - 2 | 245 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
