Assay Details
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017093.1 | NP_058789.1 | 2 - 3 | 323 | 138 | |
XM_008759109.1 | XP_008757331.1 | 2 - 3 | 310 | 138 | ||
XM_008759110.1 | XP_008757332.1 | 2 - 3 | 2587 | 138 | ||
XM_008759111.1 | XP_008757333.1 | 2 - 3 | 433 | 138 | ||
GenBank mRNA | D30041.1 | - | 2 - 3 | 323 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D30041.1, NM_017093.1, XM_008759109.1, XM_008759110.1, XM_008759111.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
