Assay Details
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017134.3 | NP_058830.2 | 1 - 2 | 207 | 63 | |
GenBank mRNA | BC091158.1 | - | 1 - 2 | 137 | 7368991 | 63 |
FM100195.1 | - | 1 - 2 | 176 | 63 | ||
FQ218591.1 | - | 1 - 2 | 151 | 63 | ||
FQ219159.1 | - | 1 - 2 | 88 | 63 | ||
J02720.1 | - | 1 - 2 | 88 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
