Assay Details
Gene Symbol: Psma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017278.1 | NP_058974.1 | 1 - 2 | 91 | 75 | |
GenBank mRNA | BC062233.1 | - | 1 - 2 | 88 | 75 | |
D90265.1 | - | 1 - 2 | 91 | 75 | ||
FQ212620.1 | - | 1 - 2 | 60 | 75 | ||
FQ213606.1 | - | 1 - 2 | 81 | 75 | ||
FQ215378.1 | - | 1 - 2 | 74 | 75 | ||
FQ218243.1 | - | 123 | 75 | |||
FQ220925.1 | - | 1 - 2 | 56 | 75 | ||
FQ222787.1 | - | 1 - 2 | 84 | 75 | ||
FQ223714.1 | - | 1 - 2 | 60 | 75 | ||
FQ229288.1 | - | 1 - 2 | 118 | 75 | ||
M29859.1 | - | 1 - 2 | 91 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC062233.1, D90265.1, M29859.1, NM_017278.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
