Assay Details
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270544.1 | NP_001257473.1 | 2 - 3 | 380 | 105 | |
NM_001270545.1 | NP_001257474.1 | 2 - 3 | 380 | 105 | ||
NM_017322.2 | NP_059018.1 | 2 - 3 | 380 | 105 | ||
XM_006246317.1 | XP_006246379.1 | 2 - 3 | 376 | 105 | ||
GenBank mRNA | BC061870.1 | - | 2 - 3 | 347 | 105 | |
L27111.1 | - | 1 - 2 | 136 | 105 | ||
L27112.1 | - | 1 - 2 | 136 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
