Assay Details
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017325.1 | NP_059021.1 | 3 - 4 | 931 | 88 | |
XM_008768598.1 | XP_008766820.1 | 3 - 4 | 2733 | 88 | ||
GenBank mRNA | L35271.1 | - | 3 - 4 | 931 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
in utero embryonic development
liver development
myeloid progenitor cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
central nervous system development
negative regulation of cell population proliferation
regulation of signal transduction
hemopoiesis
myeloid cell differentiation
neuron differentiation
negative regulation of granulocyte differentiation
positive regulation of granulocyte differentiation
hair follicle morphogenesis
response to retinoic acid
positive regulation of interleukin-2 production
embryonic hemopoiesis
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
behavioral response to pain
neuron development
definitive hemopoiesis
regulation of hair follicle cell proliferation
cellular response to transforming growth factor beta stimulus
positive regulation of cell maturation
positive regulation of progesterone secretion
transcription cis-regulatory region binding
RNA polymerase II transcription regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA-binding transcription activator activity, RNA polymerase II-specific
molecular_function
DNA binding
DNA-binding transcription factor activity
calcium ion binding
ATP binding
transcription factor binding
protein homodimerization activity
protein heterodimerization activity
DNA-binding transcription factor binding
skeletal system development
in utero embryonic development
liver development
myeloid progenitor cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
central nervous system development
negative regulation of cell population proliferation
regulation of signal transduction
hemopoiesis
myeloid cell differentiation
neuron differentiation
negative regulation of granulocyte differentiation
positive regulation of granulocyte differentiation
hair follicle morphogenesis
response to retinoic acid
positive regulation of interleukin-2 production
embryonic hemopoiesis
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
behavioral response to pain
neuron development
definitive hemopoiesis
regulation of hair follicle cell proliferation
cellular response to transforming growth factor beta stimulus
positive regulation of cell maturation
positive regulation of progesterone secretion
transcription cis-regulatory region binding
RNA polymerase II transcription regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA-binding transcription activator activity, RNA polymerase II-specific
molecular_function
DNA binding
DNA-binding transcription factor activity
calcium ion binding
ATP binding
transcription factor binding
protein homodimerization activity
protein heterodimerization activity
DNA-binding transcription factor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P228942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43480 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P228942 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P228942 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P228942 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P228942 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19449 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 139 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16130 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17905 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21395 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100059 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98993 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71114 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10753226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753229 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753225 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10753222 | - | - |
Rat Genome 230 2.0 Array | 1368914_at | - | - |
Rat Genome 230 2.0 Array | 1368914_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368914_at | - | - |
Affymetrix HT Rat Focus Array | 1368914_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100007947.rn.1 | TC1100001335.rn.1 | RTA TC |
Rat Expression Set 230 | 1368914_at | - | - |
Rat Expression Set 230 | 1368914_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753222 | - | - |
Rat Genome U34 Set | L35271_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20911630 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371943 | - | - |
RatRef-12 Expression BeadChip | ILMN_61659 | - | - |
