Assay Details
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017327.1 | NP_059023.1 | 4 - 5 | 1174 | 65 | |
XM_008772320.1 | XP_008770542.1 | 4 - 5 | 1183 | 65 | ||
XM_008772321.1 | XP_008770543.1 | 4 - 5 | 1183 | 65 | ||
XM_008772322.1 | XP_008770544.1 | 3 - 4 | 3792 | 65 | ||
GenBank mRNA | M12671.1 | - | 3 - 4 | 338 | 65 | |
M17526.1 | - | 4 - 5 | 1174 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
