Assay Details
Gene Symbol: Acox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017340.2 | NP_059036.1 | 1389 | 93 | ||
GenBank mRNA | BC085743.1 | - | 1389 | 93 | ||
J02752.1 | - | 1372 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Causes and prevention of tamoxifen-induced accumulation of triacylglycerol in rat liver.
- Authors
- Gudbrandsen OA, Rost TH, Berge RK
- Journal
- J Lipid Res - Page No 2223 (Sun Oct 01 00:00:00 UTC 2006)
- Volume (Year)
- 47 (2006)
- Product Usage
- -
