Assay Details
Gene Symbol: Gsk3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017344.1 | NP_059040.1 | 6 - 7 | 1027 | 81 | |
XM_008758972.1 | XP_008757194.1 | 6 - 7 | 1036 | 81 | ||
GenBank mRNA | X53427.1 | - | 6 - 7 | 1027 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
