Assay Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031628.1 | NP_113816.1 | 4 - 5 | 1786 | 64 | |
XM_008763702.1 | XP_008761924.1 | 5 - 6 | 3580 | 64 | ||
XM_008763703.1 | XP_008761925.1 | 4 - 5 | 3508 | 64 | ||
GenBank mRNA | D38530.1 | - | 4 - 5 | 1786 | 64 | |
DQ268830.1 | - | 4 - 5 | 1786 | 64 | ||
X86003.1 | - | 2 - 3 | 1573 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mesoderm formation
transcription, DNA-templated
transcription by RNA polymerase II
axon guidance
hippocampus development
intracellular receptor signaling pathway
adult behavior
animal organ regeneration
inner ear morphogenesis
response to hydrogen peroxide
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
response to peptide hormone
negative regulation of neuron apoptotic process
positive regulation of cell cycle
positive regulation of transcription by RNA polymerase II
semicircular canal morphogenesis
neuromuscular process controlling balance
vestibular reflex
positive regulation of leukocyte apoptotic process
DNA binding
steroid hormone receptor activity
nuclear receptor activity
zinc ion binding
sequence-specific DNA binding
mesoderm formation
transcription, DNA-templated
transcription by RNA polymerase II
axon guidance
hippocampus development
intracellular receptor signaling pathway
adult behavior
animal organ regeneration
inner ear morphogenesis
response to hydrogen peroxide
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
response to peptide hormone
negative regulation of neuron apoptotic process
positive regulation of cell cycle
positive regulation of transcription by RNA polymerase II
semicircular canal morphogenesis
neuromuscular process controlling balance
vestibular reflex
positive regulation of leukocyte apoptotic process
DNA binding
steroid hormone receptor activity
nuclear receptor activity
zinc ion binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12619 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124237 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11932 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12619 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12619 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11932 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11932 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12619 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11932 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12619 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P654444 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36791 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100706 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92718 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100758 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10868951 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868942 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868946 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868948 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868944 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868943 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868950 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868941 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868949 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868947 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868945 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10868940 | - | - |
Rat Genome 230 2.0 Array | 1393389_at | - | - |
Rat Genome 230 2.0 Array | 1369067_at | - | - |
Rat Genome 230 2.0 Array | 1369217_at | - | - |
Rat Genome 230 2.0 Array | 1393389_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369217_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369067_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393389_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369217_at | - | - |
Affymetrix HT Rat Focus Array | 1369217_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369067_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393389_at | - | - |
Affymetrix HT Rat Focus Array | 1369067_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500004149.rn.1 | TC0500000617.rn.1 | RTA TC |
Rat Expression Set 230 | 1369217_PM_at | - | - |
Rat Expression Set 230 | 1369067_at | - | - |
Rat Expression Set 230 | 1369217_at | - | - |
Rat Expression Set 230 | 1369067_PM_at | - | - |
Rat Expression Set 230 | 1393389_PM_at | - | - |
Rat Expression Set 230 | 1393389_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868940 | - | - |
Rat Genome U34 Set | rc_AI176710_at | - | - |
Rat Genome U34 Set | X86003_at | - | - |
Rat Genome U34 Set | rc_AI145834_at | - | - |
Rat Genome U34 Set | X86003_at | - | - |
Rat Genome U34 Set | rc_AI176710_at | - | - |
Rat Genome U34 Set | rc_AI145834_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21114813 | - | - |
ABI Rat Genome Survey Microarray | 21518294 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376762 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375922 | - | - |
RatRef-12 Expression BeadChip | ILMN_66003 | - | - |
