Assay Details
Gene Symbol: Rab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017355.1 | NP_059051.1 | 3 - 4 | 291 | 110 | |
GenBank mRNA | FQ227674.1 | - | 3 - 4 | 323 | 110 | |
X78605.1 | - | 3 - 4 | 291 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
