Assay Details
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017361.1 | NP_059057.1 | 5 - 6 | 785 | 86 | |
GenBank mRNA | BC078858.1 | - | 5 - 6 | 815 | 86 | |
U63840.1 | - | 5 - 6 | 785 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
