Assay Details
Gene Symbol: Tyms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019179.1 | NP_062052.1 | 1 - 2 | 187 | 77 | |
GenBank mRNA | BC126093.1 | - | 1 - 2 | 201 | 8362547 | 77 |
L12138.1 | - | 1 - 2 | 187 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
