Assay Details
Gene Symbol: Itpkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019312.2 | NP_062185.2 | 3 - 4 | 2490 | 101 | |
GenBank mRNA | AJ242781.1 | - | 2039 | 101 | ||
X74227.1 | - | 1813 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
