Assay Details
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019315.2 | NP_062188.2 | 3 - 4 | 1749 | 75 | |
GenBank mRNA | AF284345.1 | - | 3 - 4 | 1446 | 75 | |
AF292389.1 | - | 3 - 4 | 1749 | 75 | ||
U69884.1 | - | 3 - 4 | 1453 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
