Assay Details
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019315.2 | NP_062188.2 | 4 - 5 | 1890 | 72 | |
GenBank mRNA | AF284345.1 | - | 4 - 5 | 1587 | 72 | |
AF292389.1 | - | 4 - 5 | 1890 | 72 | ||
U69884.1 | - | 4 - 5 | 1594 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
