Assay Details
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019315.2 | NP_062188.2 | 7 - 8 | 2206 | 61 | |
GenBank mRNA | AF284345.1 | - | 7 - 8 | 1903 | 61 | |
AF292389.1 | - | 7 - 8 | 2206 | 61 | ||
DQ137428.1 | - | 2 - 3 | 169 | 61 | ||
U69884.1 | - | 7 - 8 | 1910 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P057594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13128 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P057594 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P057594 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P057594 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12020 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15694 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104152 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60413 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47074 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60090 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6381 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66391 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59293 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26238 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102005 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10816828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816825 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10816824 | - | - |
Rat Genome 230 2.0 Array | 1368626_at | - | - |
Rat Genome 230 2.0 Array | 1368626_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368626_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368626_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200010361.rn.1 | TC0200001585.rn.1 | RTA TC |
Rat Expression Set 230 | 1368626_at | - | - |
Rat Expression Set 230 | 1368626_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816824 | - | - |
Rat Genome U34 Set | U69884_at | - | - |
Rat Genome U34 Set | U69884_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21885397 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364332 | - | - |
RatRef-12 Expression BeadChip | ILMN_67017 | - | - |
