Assay Details
Gene Symbol: Pitx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019334.2 | NP_062207.1 | 1 - 2 | 193 | 147 | |
GenBank mRNA | AF039832.1 | - | 1 - 2 | 193 | 147 | |
AJ222972.1 | - | 1 - 2 | 246 | 148 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF039832.1, AJ222972.1, NM_019334.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
