Assay Details
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019354.3 | NP_062227.2 | 2 - 3 | 276 | 94 | |
GenBank mRNA | AB010743.1 | - | 2 - 3 | 247 | 94 | |
BC062230.1 | - | 2 - 3 | 248 | 94 | ||
FQ227097.1 | - | 2 - 3 | 276 | 94 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB010743.1, BC062230.1, NM_019354.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
- Authors
- Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
- Journal
- J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 UTC 2009)
- Volume (Year)
- 201 (2009)
- Product Usage
- -
