Assay Details
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019365.3 | NP_062238.1 | 2 - 3 | 690 | 71 | |
XM_006249796.2 | XP_006249858.1 | 2 - 3 | 1551 | 71 | ||
GenBank mRNA | AF002251.1 | - | 2 - 3 | 690 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P442802 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91285 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017298 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P442802 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017298 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P442802 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017298 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P442802 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P442802 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700540004 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13266 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74382 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36275 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36924 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17267 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700540004 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10767515 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767510 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767511 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767509 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767508 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767512 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767513 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767517 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767516 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10767507 | - | - |
Rat Genome 230 2.0 Array | 1370415_at | - | - |
Rat Genome 230 2.0 Array | 1370415_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370415_at | - | - |
Affymetrix HT Rat Focus Array | 1370415_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300009117.rn.1 | TC1300001379.rn.1 | RTA TC |
Rat Expression Set 230 | 1370415_PM_at | - | - |
Rat Expression Set 230 | 1370415_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767507 | - | - |
Rat Genome U34 Set | AF002251_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21799955 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368621 | - | - |
RatRef-12 Expression BeadChip | ILMN_56915 | - | - |
