Assay Details
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019377.1 | NP_062250.1 | 1 - 2 | 160 | 55 | |
GenBank mRNA | BC076502.1 | - | 1 - 2 | 154 | 55 | |
D17446.1 | - | 1 - 2 | 155 | 55 | ||
FQ214109.1 | - | 1 - 2 | 196 | 55 | ||
FQ229978.1 | - | 1 - 2 | 165 | 55 | ||
S55223.1 | - | 1 - 2 | 155 | 55 | ||
S83440.1 | - | 1 - 2 | 160 | 55 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC076502.1, D17446.1, NM_019377.1, S55223.1, S83440.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
